Learn More
Sino Biological Anti-ALK-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90060T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus ALK-1 / ACVRL1 ( Catalog#90060-C08H; XP_005570958.1; Met1-Gln118). ALK-1 / ACVRL1 specific IgG was purified by Cynomolgus ALK-1 / ACVRL1 affinity chromatography.
Catalog No. 50-255-2731