Learn More
Sino Biological Recombinant Cynomolgus ALK-1 Protein (ECD, His Tag)
Supplier:  Sino Biological 90060C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus ACVRL1 (XP_005570958.1) (Met1-Gln118) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2473