Learn More
Sino Biological Recombinant Anti-CD166/ALCAM Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 90227R018100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus CD166/ALCAM ( Catalog#90227-C08H; G7NZQ8; Met1-Ala526).
Catalog No. 50-240-8512