Learn More
Sino Biological Anti-CD166/ALCAM Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90227RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus CD166 / ALCAM (Catalog#90227-C08H; G7NZQ8; Met1-Ala526). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0187