Learn More
Sino Biological Cynomolgus ALCAM Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90227C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus ALCAM (G7NZQ8) (Met1-Ala526) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3595