Learn More
Sino Biological Cynomolgus ALCAM Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90227C02H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus ALCAM (NP_001244631.1) (Met1-Ala526) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-0714