Learn More
Sino Biological Cynomolgus ANGPT1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90025C07H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus ANGPT1 (XP_005563980.1) (Arg277-Phe498) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-1876