Learn More
ACROBiosystems ACROBiosystems Cynomolgus APRIL / TNFSF13 Protein, His,Flag Tag
Catalog No. 502106469
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus APRIL, His,Flag Tag (APL-C52D2) is expressed from human 293 cells (HEK293). It contains AA Gln 111 - Leu 250 (Accession # XP_005582818.1).;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 50.4 kDa.
Catalog No. 50-210-6469 Supplier ACROBiosystems Supplier No. APLC52D2100UG
Recombinant Protein;100UG;Cynomolgus APRIL, His,Flag Tag (APL-C52D2) is expressed from human 293 cells (HEK293). It contains AA Gln 111 - Leu 250 (Accession # XP_005582818.1).;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 50.4 kDa.