Learn More
Sino Biological Anti-ART1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90271T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus ART1 / CD296 / ARTC1 (Catalog#90271-C08H; XP_005579037.1; Gln23-Asn294). ART1 / CD296 / ARTC1 specific IgG was purified by Cynomolgus ART1 / CD296 / ARTC1 affinity chromatography.
Catalog No. 50-255-2689