Learn More
Sino Biological Cynomolgus ART4 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 90272C02H50
Encompass_Preferred
A DNA sequence encoding the cynomolgus ART4 (G7PJY0) (Thr31-Lys300) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2271