Learn More
ACROBiosystems ACROBiosystems Cynomolgus B7-H7 / HHLA2 Protein, His Tag (MALS verified)
Catalog No. 502106471
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus B7-H7, His Tag (B77-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Asn 345 (Accession # XP_005548285.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.2 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6471 Supplier ACROBiosystems Supplier No. B77C52H3100UG
Recombinant Protein;100UG;Cynomolgus B7-H7, His Tag (B77-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Asn 345 (Accession # XP_005548285.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.2 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.