Learn More
Sino Biological Recombinant Cynomolgus B7-H3 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 90806C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus CD276 (XP_005560056.1) (Met1-Glu465) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-228-2514