Learn More
Sino Biological Recombinant Cynomolgus VISTA/B7-H5 Protein (His Tag) 500
Supplier:  Sino Biological 90844C08H500
Encompass_Preferred
A DNA sequence encoding the cynomolgus B7H5 (XP_005565644.1) (Met1-Ala194) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6997