Learn More
ACROBiosystems ACROBiosystems Cynomolgus BAFFR / TNFRSF13C Protein, His Tag
Catalog No. 502107299
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus BAFFR, His Tag (BAR-C52H4) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Gly 76 (Accession # A0A2K5V1K1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 9.1 kDa. The protein migrates as 14-23 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) at 1 μg/mL (100 μL/well) can bind Cynomolgus BAFFR, His Tag (Cat. No. BAR-C52H4) with a linear range of 0.8-3 ng/mL (QC tested).;Immobilized Cynomolgus BAFFR, His Tag (Cat. No. BAR-C52H4) at 1 μg/mL (100 μL/well) can bind Anti-BAFFR Antibody, Human IgG1 with a linear range of 0.1-0.8 ng/mL (Routinely tested).
Catalog No. 50-210-7299 Supplier ACROBiosystems Supplier No. BARC52H4100UG
Recombinant Protein;100UG;Cynomolgus BAFFR, His Tag (BAR-C52H4) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Gly 76 (Accession # A0A2K5V1K1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 9.1 kDa. The protein migrates as 14-23 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) at 1 μg/mL (100 μL/well) can bind Cynomolgus BAFFR, His Tag (Cat. No. BAR-C52H4) with a linear range of 0.8-3 ng/mL (QC tested).;Immobilized Cynomolgus BAFFR, His Tag (Cat. No. BAR-C52H4) at 1 μg/mL (100 μL/well) can bind Anti-BAFFR Antibody, Human IgG1 with a linear range of 0.1-0.8 ng/mL (Routinely tested).