Learn More
Sino Biological Recombinant Cynomolgus CD147/Basigin/EMMPRIN Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90636C08H50
Encompass_Preferred
A DNA sequence encoding the cynomolgus CD147 (XP_005587354.1) (Met1-His209) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2674