Learn More
Sino Biological Recombinant Cynomolgus Complement component 3 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90944C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus complement C3 (XP_005587776.1) (Met1-Asn1663) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2643