Learn More
ACROBiosystems Cynomolgus CA125 25ug
Catalog No. 502535445
Encompass_Preferred
Biotinylated Cynomolgus CA125 Protein HisAvitag (CA5-C82E9) is expressed from human 293 cells (HEK293). It contains AA Pro 11544 - Pro 12188 (Accession # XP_045234963.1).
Catalog No. 50-253-5445 Supplier ACROBiosystems Supplier No. CA5C82E925UG
Biotinylated Cynomolgus CA125 Protein HisAvitag (CA5-C82E9) is expressed from human 293 cells (HEK293). It contains AA Pro 11544 - Pro 12188 (Accession # XP_045234963.1).