Learn More
Sino Biological Anti-CD14 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 90174MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Cynomolgus CD14 ( Catalog#90174-C08H; B3Y6B8; Met1-Met344). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8595