Learn More
Sino Biological Anti-CD14 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 90174MM12P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Cynomolgus CD14 ( Catalog#90174-C08H; B3Y6B8; Met1-Met344) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-0769