Learn More
Sino Biological Anti-CD14 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90174T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus CD14 ( Catalog#90174-C08H; B3Y6B8; Met1-Met344). CD14 specific IgG was purified by Cynomolgus CD14 affinity chromatography.
Catalog No. 50-241-1640