Learn More
Sino Biological Cynomolgus CD200R Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785399
Encompass_Preferred
The Cynomolgus CD200R ELISA Pair Set is for the quantitative determination of Cynomolgus CD200R.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD200R coated on a 96-well plate. Standards and samples are added to the wells, and any CD200R present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-CD200R monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of CD200R present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85399 Supplier Sino Biological Supplier No. SEK112325
The Cynomolgus CD200R ELISA Pair Set is for the quantitative determination of Cynomolgus CD200R.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD200R coated on a 96-well plate. Standards and samples are added to the wells, and any CD200R present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-CD200R monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of CD200R present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.