Learn More
Sino Biological Anti-CD200R Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11232MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Cynomolgus CD200R1 (Catalog#11232-C08H; Met 1 -Leu 267). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4765