Learn More
Sino Biological Anti-CD200R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11232RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus CD200R (rc CD200R; Catalog#11232-C08H;Met 1 -Leu 267). CD200R specific IgG was purified by cynomolgus CD200R affinity chromatography.
Catalog No. 50-255-4761