Learn More
Sino Biological Recombinant Cynomolgus CD200RLa Protein (His Tag)
Supplier:  Sino Biological 90985C08H100
Encompass_Preferred
A DNA sequence encoding the Cynomolgus CD200R1L (XP_005548211.2) (Met1-Ser240) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6570