Learn More
Sino Biological Cynomolgus CD226 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 90181C08H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus CD226 (A0A2K5W1Z6-1) (Met1-Phe252) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2165