Learn More
ACROBiosystems Cynomolgus CD3 epsilon&CD3 delta Heterodimer Protein, Fc,His Tag&Fc,Flag Tag (MALS verified)
Supplier:  ACROBiosystems CDDC52W91MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus CD3E&CD3D Heterodimer Protein, Fc,His Tag&Fc,Flag Tag (CDD-C52W9) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # Q95LI5-1 (CD3E) & Q95LI8-1 (CD3D)).;Cynomolgus CD3E&CD3D Heterodimer Protein, Fc,His Tag&Fc,Flag Tag is produced by co-expression of CD3E and CD3D, has a calculated MW of 42.5 kDa (CD3E) and 40.9 kDa (CD3D). Subunit CD3E is fused with a human IgG1 Fc tag and a polyhistidine tag at the C-terminus and subunit CD3D is fused with a human IgG1 Fc tag and a flag tag at the C-terminus. The predicted N-terminus is Gln 22 (CD3E) & Phe 22 (CD3D). The reducing (R) protein migrates as 46-50 kDa and 55-65 kDa due to glycosylation.;Immobilized Bispecific CD3×BCMA T cell-engaging Antibody at 1 μg/mL (100 μL/well) can bind Cynomolgus CD3E&CD3D Heterodimer Protein, Fc,His Tag&Fc,Flag Tag (Cat. No. CDD-C52W9) with a linear range of 1-20 ng/mL (QC tested).
Catalog No. 50-205-5094