Learn More
ACROBiosystems Biotinylated Cynomolgus CD3E&CD3D Heterodimer Protein, His,Avitag™&Tag Free (CDD-C82W6) is expressed from human 293 cells (HEK293). (Accession # Q95LI5-1 (CD3E) & Q95LI8-1 (CD3D)).
Supplier:  ACROBiosystems CDDC82W625UG
Encompass_Preferred
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Catalog No. 50-223-1027