Learn More
Sino Biological Cynomolgus CD3E Protein 1MG (Sino Biological)
Supplier:  Sino Biological 90857C02H1
Encompass_Preferred
A DNA sequence encoding the Cynomolgus CD3E (Q95LI5-1) (Met1-Asp117) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2053