Learn More
Sino Biological Recombinant Cynomolgus CD3E & CD3G Heterodimer Protein (hFc Tag)
Supplier:  Sino Biological CT091C02H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus CD3E (Q95LI5.2) (Met1-Asp117) was fused with the Fc region of human IgG1 at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the cynomolgus CD3G (NP_001270839.1) (Met1-Thr113) was fused with the Fc region of human IgG1 at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the cynomolgus CD3E/CD3G heterodimer was purified.
Catalog No. 50-255-2599