Learn More
Sino Biological Recombinant Cynomolgus CD52 Protein (hFc Tag)
Supplier:  Sino Biological 90187C02H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus CD52 (NP_001253116.1) (Met1-Ser35) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-2714