Learn More
Sino Biological Cynomolgus CD53 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90189C04H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus CD53 (G7NW46) (Glu107-Asn181) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-237-1389