Learn More
Sino Biological Cynomolgus CD58 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90190C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus CD58 (F7HQR6) (Met1-Arg215) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3027