Learn More
ACROBiosystems Cynomolgus CD7 100ug
Supplier:  ACROBiosystems CD7C52H7100UG
Encompass_Preferred
Cynomolgus CD7 Protein His Tag (CD7-C52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Ser 177 (Accession # A0A2K5VA18).
Catalog No. 50-253-5353