Learn More
ACROBiosystems Cynomolgus CD7 1mg
Catalog No. 502535352
Encompass_Preferred
Cynomolgus CD7 Protein His Tag (CD7-C52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Ser 177 (Accession # A0A2K5VA18).
Catalog No. 50-253-5352 Supplier ACROBiosystems Supplier No. CD7C52H71MG
Cynomolgus CD7 Protein His Tag (CD7-C52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Ser 177 (Accession # A0A2K5VA18).