Learn More
ACROBiosystems Cynomolgus CD7 25ug
Catalog No. 502535350
Encompass_Preferred
Biotinylated Cynomolgus CD7 Protein HisAvitag (CD7-C82E8) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Ser 177 (Accession # A0A2K5VA18).
Catalog No. 50-253-5350 Supplier ACROBiosystems Supplier No. CD7C82E825UG
Biotinylated Cynomolgus CD7 Protein HisAvitag (CD7-C82E8) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Ser 177 (Accession # A0A2K5VA18).