Learn More
Sino Biological Cynomolgus CD7 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90993H08H100
Encompass_Preferred
A DNA sequence encoding the Cynomolgus CD7 (XP_005585387.1) (Met1-Pro180) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3017