Learn More
Sino Biological Recombinant Cynomolgus CD70/CD27 Ligand Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 91021C07H1
Encompass_Preferred
A DNA sequence encoding the Cynomolgus CD70 (XP_005587773.3) (Gln39-Pro194) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-2636