Learn More
ACROBiosystems Cynomolgus CD8 alpha 50ug
Catalog No. 502535329
Encompass_Preferred
Cynomolgus CD8 alpha Protein His Tag (CDA-C52H4) is expressed from human 293 cells (HEK293). It contains AA Asn 63 - Asp 223 (Accession # XP_005575419.2).
Catalog No. 50-253-5329 Supplier ACROBiosystems Supplier No. CDAC52H450UG
Cynomolgus CD8 alpha Protein His Tag (CDA-C52H4) is expressed from human 293 cells (HEK293). It contains AA Asn 63 - Asp 223 (Accession # XP_005575419.2).