Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Cynomolgus CD8 Alpha & Beta Heterodimer Protein, His-Tagged (Lyophilized, 100 μg)

Catalog No. NC3779718
Encompass_Preferred

Recombinant cynomolgus CD8 alpha and beta heterodimer expressed in HEK293 cells. CD8 alpha contains a C-terminal polyhistidine tag and CD8 beta is untagged. The protein is supplied lyophilized, verified by SEC-MALS for heterodimer assembly, and suitable for antibody binding and biochemical characterization.

  • Species coverage: Cynomolgus macaque
  • Expression system: Human 293 cells (HEK293)
  • Tag: CD8 alpha with C-terminal His tag; CD8 beta untagged
  • Purity: >95% by SDS-PAGE; >85% by SEC-MALS
  • Molecular weight: Calculated 23.7 kDa (alpha), 21.1 kDa (beta); apparent 30-35 kDa on SDS-PAGE
  • Formulation: Lyophilized from PBS, pH 7.4, with trehalose
  • Endotoxin: <1.0 EU/μg
  • Storage stability: 12 months lyophilized at -20°C to -70°C

Catalog No. NC3779718 Supplier ACROBiosystems Supplier No. CDAC52W3100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant cynomolgus CD8 alpha and beta heterodimer expressed in HEK293 cells. CD8 alpha contains a C-terminal polyhistidine tag and CD8 beta is untagged. The protein is supplied lyophilized, verified by SEC-MALS for heterodimer assembly, and suitable for antibody binding and biochemical characterization.

  • Species coverage: Cynomolgus macaque
  • Expression system: Human 293 cells (HEK293)
  • Tag: CD8 alpha with C-terminal His tag; CD8 beta untagged
  • Purity: >95% by SDS-PAGE; >85% by SEC-MALS
  • Molecular weight: Calculated 23.7 kDa (alpha), 21.1 kDa (beta); apparent 30-35 kDa on SDS-PAGE
  • Formulation: Lyophilized from PBS, pH 7.4, with trehalose
  • Endotoxin: <1.0 EU/μg
  • Storage stability: 12 months lyophilized at -20°C to -70°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.