Learn More
Sino Biological Anti-CD83 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90199RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus / Rhesus CD83 (Catalog#90199-C08H; F6WX37; Met1-Ala143). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2712