Learn More
Sino Biological Recombinant Cynomolgus CD8A & CD8B Heterodimer Protein (Flag & His Tag)
Supplier:  Sino Biological CT077C2508H50
Encompass_Preferred
A DNA sequence encoding the cynomolgus CD8A (XP_005575419.1) (Met42-Asp223) was fused with a flag tag at the C-terminus, constructed the plasmid 1 ; A DNA sequence encoding the cynomolgus CD8B (XP_005575417.1) (Met1-Pro170) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the heterodimer was purified.
Catalog No. 50-224-6690