Learn More
Sino Biological Recombinant Cynomolgus CD8 alpha Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 90888C02H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus CD8A (XP_005575419.1) (Met42-Asp223) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3122