Learn More
ACROBiosystems Cynomolgus CDCP1 Protein, His Tag (MALS verified)
Catalog No. 502719844
Encompass_Preferred
CUB domain-containing protein 1, also know as CDCP1. CDCP1 is a cell surface glycoprotein that has been recognized both as a tumour marker and as a potential target to disrupt progression of cancer. CDCP1 has been shown to modulate cellsubstratum adhesion and motility in colon cancer cell lines.
Catalog No. 50-271-9844 Supplier ACROBiosystems Supplier No. CD1C52H31MG
CUB domain-containing protein 1, also know as CDCP1. CDCP1 is a cell surface glycoprotein that has been recognized both as a tumour marker and as a potential target to disrupt progression of cancer. CDCP1 has been shown to modulate cellsubstratum adhesion and motility in colon cancer cell lines.