Learn More
Sino Biological Recombinant Cynomolgus Cadherin-17 Protein (mFc Tag), HPLC-verified
Catalog No. 502992745
Encompass_Preferred
A DNA sequence encoding the Cynomolgus CDH17 (XP_005563762.2) (Met1-Thr784) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-299-2745 Supplier Sino Biological Supplier No. 90994C38H1
A DNA sequence encoding the Cynomolgus CDH17 (XP_005563762.2) (Met1-Thr784) was expressed with the Fc region of mouse IgG2a at the C-terminus.