Learn More
Sino Biological Recombinant Cynomolgus CEACAM-5/CD66e Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 90891C49HB100
Encompass_Preferred
A DNA sequence encoding the Cynomolgus CEACAM5 (XP_005589491.1) (Met1-Ala688) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-1202