Learn More
Sino Biological Recombinant Cynomolgus CEACAM-5/CD66e Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 90891C02H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus CEACAM5 (XP_005589491.1) (Met1-Ala688) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-228-2481