Learn More
Sino Biological Recombinant Cynomolgus CEACAM-6/CD66c Protein (His Tag), HPLC-verified 500
Supplier:  Sino Biological 90940C08H500
Encompass_Preferred
A DNA sequence encoding the cynomolgus CEACAM6 (XP_005589488.1) (Met1-Pro323) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6988