Learn More
ACROBiosystems ACROBiosystems Cynomolgus Coagulation Factor III / Tissue Factor Protein, His Tag (active enzyme, MALS verified)
Catalog No. 502106256
Encompass_Preferred
Recombinant Protein;500UG;Cynomolgus Tissue Factor, His Tag (TF3-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Glu 252 (Accession # A0A2K5VX02-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.9 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus Tissue Factor, His Tag (Cat. No. TF3-C52H3) at 1 μg/mL (100 μL/well) can bind Anti-TF Antibody, Human IgG1 with a linear range of 0.2-3 ng/mL (Routinely tested).
Catalog No. 50-210-6256 Supplier ACROBiosystems Supplier No. TF3C52H3500UG
Recombinant Protein;500UG;Cynomolgus Tissue Factor, His Tag (TF3-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Glu 252 (Accession # A0A2K5VX02-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.9 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus Tissue Factor, His Tag (Cat. No. TF3-C52H3) at 1 μg/mL (100 μL/well) can bind Anti-TF Antibody, Human IgG1 with a linear range of 0.2-3 ng/mL (Routinely tested).