Learn More
ACROBiosystems Cynomolgus Complement C2 200ug
Catalog No. 502535233
Encompass_Preferred
Biotinylated Cynomolgus Complement C2 Protein HisAvitag (CO2-C82E7) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Leu 752 (Accession # G8F3W0).
Catalog No. 50-253-5233 Supplier ACROBiosystems Supplier No. CO2C82E7200UG
Biotinylated Cynomolgus Complement C2 Protein HisAvitag (CO2-C82E7) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Leu 752 (Accession # G8F3W0).