Learn More
ACROBiosystems Cynomolgus Complement C2 25ug
Supplier:  ACROBiosystems CO2C82E725UG
Encompass_Preferred
Biotinylated Cynomolgus Complement C2 Protein HisAvitag (CO2-C82E7) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Leu 752 (Accession # G8F3W0).
Catalog No. 50-253-5232